Learn More
Grainger Block 6 X 50Ml
Catalog No. 502752651
Encompass_Preferred
The Grainger 14N145 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, extension, or other accessory. It is rated up to 20,000 psi torque and measures 1-1/2 inches long. The Westward 14N145 provides a simple way to increase the versatility of your 1/4-inch drive tools.
Catalog No. 50-275-2651 Supplier Grainger Supplier No. 14N145
The Grainger 14N145 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, extension, or other accessory. It is rated up to 20,000 psi torque and measures 1-1/2 inches long. The Westward 14N145 provides a simple way to increase the versatility of your 1/4-inch drive tools.