Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for 6 mm Test Tubes, 30 Well, 2070Q
SDP
Catalog No. 5032290
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 30 Well
Model: 2070Q
Opening: 0.31"(.79 cm)
Catalog No. 50-322-90 Supplier Electron Microscopy Sciences Supplier No. 6312901
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 30 Well
Model: 2070Q
Opening: 0.31"(.79 cm)