Learn More
Grainger Block 96 X 0.2Ml Or One Pcr Plate
Catalog No. 502751782
Encompass_Preferred
The Grainger Approved 96-Well 0.2 mL PCR Plate is made of clear polypropylene for visibility. It has 96 wells arranged in an 8x12 format for PCR reactions. The wells have a maximum volume of 200 μL. The plates have alphanumeric grid reference for easy sample tracking. The plates are free of DNase, RNase and human genomic DNA. They are non-sterile and disposable. The dimensions are 127 x 85 x 14 mm. These PCR plates are suitable for thermal cycling.
Catalog No. 50-275-1782 Supplier Grainger Supplier No. 14N138
The Grainger Approved 96-Well 0.2 mL PCR Plate is made of clear polypropylene for visibility. It has 96 wells arranged in an 8x12 format for PCR reactions. The wells have a maximum volume of 200 μL. The plates have alphanumeric grid reference for easy sample tracking. The plates are free of DNase, RNase and human genomic DNA. They are non-sterile and disposable. The dimensions are 127 x 85 x 14 mm. These PCR plates are suitable for thermal cycling.