Learn More
AbD Serotec Block ACE, 20 X 4g
Supplier:  AbD Serotec BUF029
Encompass
Block Ace is designed as a high-performance blocking reagent for use in immunological assays such as ELISA and Western blotting. It may also be used for dilution of antibodies and for washing procedures in the above assays.
Catalog No. NC1588655