Learn More
Newcomer Supply Tissue Block Storage System, Plastic - CassCab 6, Six Drawer Unit that Stores up to 1,500 Tissue Block Cassettes!
SDP
Catalog No. NC2101881
Encompass
The block storage system has six segmented, impact resistant plastic drawers with ID area per unit. There are seven segmented rows per drawer and each row includes a foam block to support blocks in upright position. The cabinets have locking pegs at each corner for stacking multiple units on one another. Each 6 drawer unit accommodates up to 1,500 blocks or 1,000 rings. Dimensions: 17 3/8" (L) x 9 1/2" (W) x 15 1/4" (H).
This item is not returnable. View return policy
Catalog No. NC2101881 Supplier Newcomer Supply Supplier No. 6883
The block storage system has six segmented, impact resistant plastic drawers with ID area per unit. There are seven segmented rows per drawer and each row includes a foam block to support blocks in upright position. The cabinets have locking pegs at each corner for stacking multiple units on one another. Each 6 drawer unit accommodates up to 1,500 blocks or 1,000 rings. Dimensions: 17 3/8" (L) x 9 1/2" (W) x 15 1/4" (H).