Learn More
ROSE SCIENTIFIC INC HANON Kjeldahl Block Digestor, Graphite System SH220F, 220 V, 50/60Hz
Catalog No. NC3666727
Encompass
(Available only in 220V, 50/60Hz)Suitable for 20 x 300mL digestion tubesNetwWeight: 25 KgDimensions: 515x458x730mm
Catalog No. NC3666727 Supplier ROSE SCIENTIFIC INC Supplier No. B0100000129
(Available only in 220V, 50/60Hz)Suitable for 20 x 300mL digestion tubesNetwWeight: 25 KgDimensions: 515x458x730mm