Learn More
MSC Block Division Pulley Block, 2.5 in Double Sheave, 685 lb Load
Catalog No. 5000528122
Encompass
This industrial-quality swivel eye pulley block features a 2.5 in double machined sheave with an oil-impregnated bushing, grooved for 1/4 in wire ropes. It has a clear zinc finish and a safe working load of 685 lb.
  • Swivel eye design for versatile rigging.
  • Double machined sheave for efficient operation.
  • Oil impregnated bushing for smooth rotation.
  • Customizable with bolt fasteners or a snatch pin.
Catalog No. 50-005-28122 Supplier MSC Supplier No. 67326389
This industrial-quality swivel eye pulley block features a 2.5 in double machined sheave with an oil-impregnated bushing, grooved for 1/4 in wire ropes. It has a clear zinc finish and a safe working load of 685 lb.
  • Swivel eye design for versatile rigging.
  • Double machined sheave for efficient operation.
  • Oil impregnated bushing for smooth rotation.
  • Customizable with bolt fasteners or a snatch pin.