Learn More
Grainger Block For One Micro Plate
Catalog No. 502751655
Encompass_Preferred
The Block For One Micro Plate by Block Scientific is a 96-well microplate storage block made of polypropylene that can hold one SBS footprint microplate. It features alphanumeric grid reference for easy sample location. This storage block measures 127 x 86 x 45 mm (5 x 3.4 x 1.8 inches) and weighs 0.1 kg (0.22 lb). It can withstand temperatures from -20°C to 121°C (-4°F to 250°F) and is autoclavable. This polypropylene microplate block provides an organized storage solution for 96-well microplates.
Catalog No. 50-275-1655 Supplier Grainger Supplier No. 14N146
The Block For One Micro Plate by Block Scientific is a 96-well microplate storage block made of polypropylene that can hold one SBS footprint microplate. It features alphanumeric grid reference for easy sample location. This storage block measures 127 x 86 x 45 mm (5 x 3.4 x 1.8 inches) and weighs 0.1 kg (0.22 lb). It can withstand temperatures from -20°C to 121°C (-4°F to 250°F) and is autoclavable. This polypropylene microplate block provides an organized storage solution for 96-well microplates.