Learn More
Cole-Parmer Stuart Digital 2-Block Heater, 200⁰C, 120 V
Supplier:  Cole-Parmer 3661020
Encompass
Stuart dry block heaters are designed for precise heating of test tubes, microcentrifuge tubes, cuvettes, and microtiter plates. The "number of blocks accepted" indicates the number of single-width blocks accepted by each heater. A heater that accepts two single-width blocks will also accept one double-width block.
Catalog No. 50-196-0432