Learn More
Cole-Parmer Techne Dri-Block Digital Block Heater, Dual Insert, 200C; 230 VAC
Catalog No. 501960484 Shop All Cole-Parmer Products
Encompass
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out. The "number of blocks accepted" on the ordering table indicate the number of single-width blocks accepted by each block heater. A heater that accepts two single-width blocks will accept one double-width block. Use three- and four-block models for high-throughput applications such as chemical oxygen demand (COD).
Catalog No. 50-196-0484 Supplier Cole-Parmer Supplier No. 3662008
Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out. The "number of blocks accepted" on the ordering table indicate the number of single-width blocks accepted by each block heater. A heater that accepts two single-width blocks will accept one double-width block. Use three- and four-block models for high-throughput applications such as chemical oxygen demand (COD).