Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control

Catalog No. 14750100
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 125 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-750-100 2 x 125 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-750-100 Supplier Invitrogen™ Supplier No. 14750100
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control

The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is an ideal indicator of lipid-mediated transfection efficiency for RNAi experiments, particularly those using the Lipofectamine™ RNAiMAX Transfection Reagent. The sequence of the BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is not homologous to any known gene, preventing induction of nonspecific cellular events caused by introduction of the duplex into cells. The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control provides:
  • A strong, easily detectable, and photostable Alexa Fluor™ 555* signal indicating transfection efficiency
  • Chemical modifications for a clearer, more persistent cellular signal than with labeled siRNA
  • Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules *This dye can be detected using standard filter sets designed for Alexa Fluor™ 555, Cy™3, DsRed, Texas Red™, or rhodamine fluorophores.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The BLOCK-iT™ Alexa Fluor™ Red Fluorescent Control is provided at a concentration of 20 μM. Store at -20°C. Guaranteed stable for 12 months when properly stored.
    Control Type Transfection Control
    Detection Method Fluorescence
    Sample Type RNA
    For Use With (Application) RNA Transfection
    Label or Dye Alexa Fluor 555
    Product Line Alexa Fluor, BLOCK-iT
    Product Type Red fluorescent transfection control
    Quantity 2 x 125 μL
    RNAi Type siRNA
    Shipping Condition Dry Ice
    Show More Show Less
    What are the positive and negative controls I should use with my Stealth RNAi siRNA experiment?

    We offer several negative and positive controls for Stealth siRNA:

    For Stealth Positive Control:
    - Stealth RNAi siRNA GAPDH Positive Control (human) (Cat. No. 12935140, 250 µL)
    - Stealth RNAi siRNA beta-Lactamase Reporter Control (Cat. No. 12935148, 250 µL)
    - Stealth RNAi siRNA GFP Reporter Control (Cat. No. 12935145, 250 µL)
    - Stealth RNAi siRNA Luciferase Reporter Control (Cat. No. 12935146, 250 µL)

    For Stealth Negative Control:
    Hi (55-65% GC), Med (45-55% GC), or Low (35-45% GC) Duplexes are offered:

    - Stealth RNAi siRNA Negative Control Hi GC (Cat. No. 12935400, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #2 (Cat. No. 12935114, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935115, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex (Cat. No. 12935300, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex #2 (Cat. No. 12935112, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935113, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex (Cat. No. 12935200, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #2 (Cat. No. 12935110, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #3 (Cat. No. 12935111, 250 µL)
    - Stealth RNAi siRNA Negative Control Kit (Cat. No. 12935100, contains one of each GC content level negative control, 250 µL each at 20 µM)
    - For a scrambled negative control, you could also scramble your designed siRNA or Stealth RNAi sequence using the BLOCK-iT RNAi Designer.

    You can also use our BLOCK-iT AlexaFluor Red Fluorescent Control (Cat. No. 14750100) or BLOCK-iT Fluorescent Oligo (Cat. No. 2013, 13750062) as a control for the transfection.

    Can I use the BLOCK-iT Fluorescent Oligo, for lipid transfection (Cat. No. 2013) to assess transfection efficiency with Lipofectamine RNAiMAX Transfection Reagent (Cat. No. 13778100)?

  • We do not recommend using BLOCK-iT Fluorescent Oligo, for lipid transfection (Cat. No. 2013) to assess transfection efficiency with Lipofectamine RNAiMAX Transfection Reagent (Cat. No. 13778100). BLOCK-iT Alexa Fluor Red Fluorescent Control (Cat. No. 14750100) should be used to assess transfection efficiency with Lipofectamine RNAiMAX Reagent.
  • BLOCK-iT Fluorescent Oligo, for lipid transfection is optimized for use with Lipofectamine 2000 Transfection Reagent and is not recommended for Lipofectamine RNAiMAX Reagent.
  • For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.