Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BLOCK-iT™ Fluorescent Oligo, for electroporation

Catalog No. 13750062
Encompass_Preferred
Change view
Click to view available options
Quantity:
75 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-750-062 75 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-750-062 Supplier Invitrogen™ Supplier No. 13750062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BLOCK-iT™ Fluorescent Oligo, for electroporation

BLOCK-iT™ Fluorescent Oligo provides:

  • A strong, easily detectable FITC signal indicating transfection efficiency
  • Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA
  • Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules

    This makes the BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. BLOCK-iT™ Fluorescent Oligo is available in two configurations: one designed for lipid-mediated transfection (2 x 125μL) at 20μM and one designed for electro-poration (75μl) at 1 mM.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The BLOCK-iT™ Fluorescent Oligo for electroporation is supplied as 75 μl at 1 mM, and should be stored at -20° C.
    Control Type Transfection Control
    Detection Method Fluorescence
    Sample Type RNA
    For Use With (Application) Electroporation
    Label or Dye FITC (Fluorescein)
    Product Line BLOCK-iT
    Product Type Fluorescent Oligo
    Quantity 75 μL
    RNAi Type siRNA
    What are the positive and negative controls I should use with my Stealth RNAi siRNA experiment?

    We offer several negative and positive controls for Stealth siRNA:

    For Stealth Positive Control:
    - Stealth RNAi siRNA GAPDH Positive Control (human) (Cat. No. 12935140, 250 µL)
    - Stealth RNAi siRNA beta-Lactamase Reporter Control (Cat. No. 12935148, 250 µL)
    - Stealth RNAi siRNA GFP Reporter Control (Cat. No. 12935145, 250 µL)
    - Stealth RNAi siRNA Luciferase Reporter Control (Cat. No. 12935146, 250 µL)

    For Stealth Negative Control:
    Hi (55-65% GC), Med (45-55% GC), or Low (35-45% GC) Duplexes are offered:

    - Stealth RNAi siRNA Negative Control Hi GC (Cat. No. 12935400, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #2 (Cat. No. 12935114, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935115, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex (Cat. No. 12935300, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex #2 (Cat. No. 12935112, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935113, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex (Cat. No. 12935200, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #2 (Cat. No. 12935110, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #3 (Cat. No. 12935111, 250 µL)
    - Stealth RNAi siRNA Negative Control Kit (Cat. No. 12935100, contains one of each GC content level negative control, 250 µL each at 20 µM)
    - For a scrambled negative control, you could also scramble your designed siRNA or Stealth RNAi sequence using the BLOCK-iT RNAi Designer.

    You can also use our BLOCK-iT AlexaFluor Red Fluorescent Control (Cat. No. 14750100) or BLOCK-iT Fluorescent Oligo (Cat. No. 2013, 13750062) as a control for the transfection.

    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.