Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for 0.2 ml Microcentrifuge Tube, 79 Wells, 2059Q
SDP
Catalog No. 5032277
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 79 Wells
Model: 2059Q
Opening: 0.24"(0.61 cm) taper
Catalog No. 50-322-77 Supplier Electron Microscopy Sciences Supplier No. 6313102
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 79 Wells
Model: 2059Q
Opening: 0.24"(0.61 cm) taper