Learn More
Newcomer Supply Tissue Block Modular Storage Drawers, Plastic, 6 drawers/case. Each drawer holds up to 250 tissue blocks
SDP
Catalog No. NC1887037
Encompass
The plastic Tissue Block Modular Storage Drawers provide permanent storage and identification of embedding cassettes and rings. Each drawer is made of high impact resistant plastic and holds up to 250 cassettes or 165 embedding rings. The drawers are stackable with interlocking ridges on top and bottom. Dimensions: 15 7/8" x 9 1/8" x 2 1/8". Identification labels included.
Catalog No. NC1887037 Supplier Newcomer Supply Supplier No. 6884
The plastic Tissue Block Modular Storage Drawers provide permanent storage and identification of embedding cassettes and rings. Each drawer is made of high impact resistant plastic and holds up to 250 cassettes or 165 embedding rings. The drawers are stackable with interlocking ridges on top and bottom. Dimensions: 15 7/8" x 9 1/8" x 2 1/8". Identification labels included.