Learn More
Chemglass Life Sciences MultiTherm Block, 12 x 15mL (200 to 750rpm, Ambient -10° to 100°C)
SDP
Catalog No. 50143449
Encompass
MultiTherm Block, 12 x 15mL (200 to 750rpm, Ambient -10¿ to 100¿C).
Catalog No. 50-143-449 Supplier Chemglass Life Sciences Supplier No. CLS4030017
MultiTherm Block, 12 x 15mL (200 to 750rpm, Ambient -10¿ to 100¿C).