Learn More
Chemglass Life Sciences MultiTherm Block, 6 x 50mL (200 to 750rpm, Ambient -10° to 100°C)
SDP
Catalog No. 50143450
Encompass
MultiTherm Block, 6 x 50mL (200 to 750rpm, Ambient -10¿ to 100¿C).
Catalog No. 50-143-450 Supplier Chemglass Life Sciences Supplier No. CLS4030019
MultiTherm Block, 6 x 50mL (200 to 750rpm, Ambient -10¿ to 100¿C).