Learn More
IMEB, Inc Base (Only) forTek-Select Paraffin Block or Microscope Slide Metal Storage Cabinet 14 Drawers, Stackable.
SDP
Catalog No. NC2598177
Encompass_Preferred
Base (Only) forTek-Select Paraffin Block or Microscope Slide Metal Storage Cabinet 14 Drawers, Stackable. Tek-Select economical light weight metal microscope slide and paraffin block storage cabinets allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15″ high. Cassette Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers.
Catalog No. NC2598177 Supplier IMEB, Inc Supplier No. SC010
Base (Only) forTek-Select Paraffin Block or Microscope Slide Metal Storage Cabinet 14 Drawers, Stackable. Tek-Select economical light weight metal microscope slide and paraffin block storage cabinets allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15″ high. Cassette Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers.