Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Block-Out Base Mold Release
SDP

Catalog No. 22000015
Change view
Click to view available options
150 mL 1EA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-000-015 150 mL 1EA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-015 Supplier StatLab Supplier No. BMR4
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

An excellent base mold release compound

  • Designed to be used as a base mold coating to allow for easy separation of paraffin from the base mold
  • Helps to prevent paraffin build-up on metal base molds
  • 150 mL bottle

Specifications

Physical Form Solution
CAS 64-17-5, 67-63-0
Color Clear Colorless
Quantity 150 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.