Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for PCR Block, size of two blocks, 2083Q
SDP
Catalog No. 5032281
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Model: 2083Q
Catalog No. 50-322-81 Supplier Electron Microscopy Sciences Supplier No. 6313250
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Model: 2083Q