Learn More
Grainger Block Pcr Plate Skirted/Non-Skirted
Catalog No. 502749632
Encompass_Preferred
The Grainger 14N152 is a 14 inch diameter, 3 blade, right hand rotation, cast aluminum fan blade. It has a 10 degree pitch and is balanced for optimal airflow. The blades are unfinished and unpainted. This is a replacement blade for exhaust fans, cooling fans, circulator fans, and ventilation fans. It can be used with fan motors up to 1/2 HP and fits a 5/8 inch diameter shaft. The blade measures 14 inches across and is 1 inch thick. It is a medium duty blade made in the USA out of sturdy aluminum.
Catalog No. 50-274-9632 Supplier Grainger Supplier No. 14N152
The Grainger 14N152 is a 14 inch diameter, 3 blade, right hand rotation, cast aluminum fan blade. It has a 10 degree pitch and is balanced for optimal airflow. The blades are unfinished and unpainted. This is a replacement blade for exhaust fans, cooling fans, circulator fans, and ventilation fans. It can be used with fan motors up to 1/2 HP and fits a 5/8 inch diameter shaft. The blade measures 14 inches across and is 1 inch thick. It is a medium duty blade made in the USA out of sturdy aluminum.