Learn More
Enzo Life Sciences Blocking peptide for Glutamic acid decarboxylase mAb (11) (Prod. No. BML-GC3208) (50µg)
SDP
Supplier:  Enzo Life Sciences BMLGP32090050
Encompass_Preferred
Immunization and blocking peptide for Glutamic Acid Decarboxylase (GAD) antibody (BML-GC3208). The tetradecapeptide corresponds to the N-terminal residues 4-17 of rat GAD65. The sequence is wholly conserved human GAD65. Purity: ≥99%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-0910