Learn More
Enzo Life Sciences Blocking peptide for Glutamic acid decarboxylase pAb (Prod. No. BML-GC3008) (50µg)
SDP
Supplier:  Enzo Life Sciences BMLGP30090050
Encompass_Preferred
Immunization and blocking peptide for Glutamic Acid Decarboxylase (GAD) antibodies. The pentadecapeptide corresponds to the C-terminal residues [Cys+]572-585 of rat GAD65. The sequence is wholly conserved at the C-terminus of rat GAD67, mouse, pig and human GAD65/67, cat GAD67, and in human GAD-2. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-0909