Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for Stainless steel cover for models 2064Q and 2065Q
SDP
Catalog No. 5032274
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Catalog No. 50-322-74 Supplier Electron Microscopy Sciences Supplier No. 63134
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.