Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for Titer Plate, size of two blocks, 1 Well, 2065Q
SDP
Supplier:  Electron Microscopy Sciences 6313302
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 1 Well
Model: 2065Q
Catalog No. 50-322-84