Learn More
Grainger Block Tubes 60Mm Tall No Lid
Catalog No. 502752798
Encompass_Preferred
The Benchmark Scientific Aluminum Block for Tubes is a block made of aluminum that holds test tubes and vials. It has holes drilled in it to accommodate test tubes and vials of various sizes including 13mm, 16mm, 18mm, and 25mm. The block allows for convenient organization and storage of multiple tubes and vials in one place and provides a stable base to hold them upright. The aluminum construction makes it durable and resistant to corrosion. This tube rack is a useful benchtop accessory for any laboratory setting.
Catalog No. 50-275-2798 Supplier Grainger Supplier No. 14N148
The Benchmark Scientific Aluminum Block for Tubes is a block made of aluminum that holds test tubes and vials. It has holes drilled in it to accommodate test tubes and vials of various sizes including 13mm, 16mm, 18mm, and 25mm. The block allows for convenient organization and storage of multiple tubes and vials in one place and provides a stable base to hold them upright. The aluminum construction makes it durable and resistant to corrosion. This tube rack is a useful benchtop accessory for any laboratory setting.