Learn More
Mitegen Block, Wizard PEG Ion 8K and 10K, 96 formulations, 1.7 mL volumes
SDP
Catalog No. 501492090
Encompass_Preferred
The PEG Ion series matrix combines the crystallizing power of polyethylene glycols (PEGs) with salts that are commonly found in crystallization conditions. Use PEG Ion during the coarse screening phase of biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).96-Well Block Plate formulations are 1.7 ml each in a 96 deep-well matrix block plate. All formulations are prepared with ASTM type-1 water and high-purity chemicals, 0.2 µm filtered in sterile packaging.
Catalog No. 50-149-2090 Supplier Mitegen Supplier No. 1008656
Add to Cart
Add to Cart
The PEG Ion series matrix combines the crystallizing power of polyethylene glycols (PEGs) with salts that are commonly found in crystallization conditions. Use PEG Ion during the coarse screening phase of biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).96-Well Block Plate formulations are 1.7 ml each in a 96 deep-well matrix block plate. All formulations are prepared with ASTM type-1 water and high-purity chemicals, 0.2 µm filtered in sterile packaging.