Learn More
Mitegen Block, Wizard JCSG Plus, 96 formulations, 1.7 mL volumes
SDP
Catalog No. 501492089
Encompass_Preferred
The Wizard JCSG+™ 96 well block plate sparse matrix protein crystallization screen is designed for coarse screening of initial crystallization and solubility conditions for biological macromolecules. Researchers from the Joint Center for Structural Genomics (JCSG) formulated this screen with the goal of maximizing the coverage of the crystallization parameter space (buffers, salts, precipitants, and pH) and eliminating redundancy. The buffers give final solution pHs between 4.2 and 10.5. Precipitants include volatile reagents such as ethanol, high molecular weight polymers such as PEG-8000, and salts such as ammonium sulfate.
Catalog No. 50-149-2089 Supplier Mitegen Supplier No. 1008651
Add to Cart
Add to Cart
The Wizard JCSG+™ 96 well block plate sparse matrix protein crystallization screen is designed for coarse screening of initial crystallization and solubility conditions for biological macromolecules. Researchers from the Joint Center for Structural Genomics (JCSG) formulated this screen with the goal of maximizing the coverage of the crystallization parameter space (buffers, salts, precipitants, and pH) and eliminating redundancy. The buffers give final solution pHs between 4.2 and 10.5. Precipitants include volatile reagents such as ethanol, high molecular weight polymers such as PEG-8000, and salts such as ammonium sulfate.