Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Blockade Versatile Sign Stand

Catalog No. 19169612 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Galvanized Steel, Cast Iron
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-169-612 Galvanized Steel, Cast Iron
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-612 Supplier Accuform™ Supplier No. HSP820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blockade Versatile Sign Stand is ideal for quickly and easily posting signs indoors or outside.

  • Base is solid, heavyweight cast iron material, weighing 37 lb.
  • Powder-coated in black and gray
  • Foldable legs flip open to increase footprint from 18 to 36 in. for added stability
  • Legs have a quick and easy locking feature to hold them in place
  • Includes a 6 ft. x 1.75 in. square galvanized steel post

Specifications

Material Galvanized Steel, Cast Iron
Mounting Type Floor-standing
Height (English) 6 ft.
Product Type Versatile Sign Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.