Description
- Collapsible for easier transport
- Fits cell phones, tablets, and similarly sized devices
- Includes one carrying case
- Built-in features:
- One Double-layer conductive glove
- One large transparent window
- Two accessory pockets
- At least two layers of High-shielding Faraday fabric on all sides
- Dual-paired seam construction
- Unique serial number for asset tracking
- Multi-layer shielding fabric
- Materials Includes:
- Ballistic nylon
- High-shielding Faraday fabric
- Water-resistant outer materials
- Padded interior walls
- Dimensions Includes:
- Internal: 20 x 15.5 x 11.5 in.
- Folds down to: 14 x 10 x 2 in.
- Designed for law enforcement and military forensic investigators to analyze devices while shielded
- Offers space for accessories such as batteries and data extraction tools
- Allows users to operate and view touchscreen devices while maintaining shielding
- Isolates devices and blocks Wi-Fi (2.4 and 5 GHz), Bluetooth, cellular, GPS, RFID, and radio signals (60-80 dB)
Specifications
Specifications
| Product Type | Faraday Bag |