Learn More
Active Motif Blocker
SDP
Catalog No. NC1847620
Encompass
Reduce the effects of non-specific binding in your chromatin immunoprecipitation (ChIP) reactions by including Blocker.
Catalog No. NC1847620 Supplier Active Motif Supplier No. 37498
Add to Cart
Add to Cart
Reduce the effects of non-specific binding in your chromatin immunoprecipitation (ChIP) reactions by including Blocker.