Learn More
Rockland Immunochemicals Blocking Buffer for Fluorescent Western Blotting 10-PACK (10 x 500 ml) - MB-070-010
SDP
Catalog No. NC9365641
Encompass_Preferred
Western Blot Blocking Buffer is specifically designed for western blotting using fluorochrome conjugated antibodies. Pure nitrocellulose membrane is recommended for maximum performance. Other membranes, such as PVDF or nitrocellulose embedded in a support can be used, but may generate elevated backgrounds. Protein should be transferred from gel to membrane using standard protocols. This buffer can be used for membrane blocking and to dilute both primary and secondary antibodies. See www.rockland-inc.com for specific protocols. This product is suitable for use with fluorescent western blot imaging systems produced by Bio-Rad Laboratories, GE Healthcare, Alpha Innotech, FujiFilm Life Science, Licor Biosciences, UVP and Syngene.
Catalog No. NC9365641 Supplier Rockland Immunochemicals Supplier No. MB070010
Western Blot Blocking Buffer is specifically designed for western blotting using fluorochrome conjugated antibodies. Pure nitrocellulose membrane is recommended for maximum performance. Other membranes, such as PVDF or nitrocellulose embedded in a support can be used, but may generate elevated backgrounds. Protein should be transferred from gel to membrane using standard protocols. This buffer can be used for membrane blocking and to dilute both primary and secondary antibodies. See www.rockland-inc.com for specific protocols. This product is suitable for use with fluorescent western blot imaging systems produced by Bio-Rad Laboratories, GE Healthcare, Alpha Innotech, FujiFilm Life Science, Licor Biosciences, UVP and Syngene.