Learn More
Biotium Blocking Buffers Background Reducers Fluorescence Microscopy Chemical Reagents TrueBlack 1/EA
Catalog No. 5000501533
Encompass_Preferred
This buffer is a component of the TrueBlack Background Suppressor System (Permeabilizing) It is an excellent general use non-mammalian blocking buffer with detergent for intracellular immunofluorescence Not recommended for use with HRP (contains azide) 100 mL bottle
Catalog No. 50-005-01533 Supplier Biotium Supplier No. 23012B100ML
This buffer is a component of the TrueBlack Background Suppressor System (Permeabilizing) It is an excellent general use non-mammalian blocking buffer with detergent for intracellular immunofluorescence Not recommended for use with HRP (contains azide) 100 mL bottle