Learn More
Biotium Blocking Buffers Background Reducers Fluorescence Microscopy Western and Protein Analysis Chemical Reagents 1/EA
Catalog No. 5000501479
Encompass_Preferred
This buffer is a component of the TrueBlack WB Blocking Kit It is available for customers who have evaluated the kit and prefer to use the Antibody Diluent for membrane blocking and antibody incubation steps For blocking and antibody dilution for 50 membranes (1 L buffer 10 mL per membrane)
Catalog No. 50-005-01479 Supplier Biotium Supplier No. 23013B1L
This buffer is a component of the TrueBlack WB Blocking Kit It is available for customers who have evaluated the kit and prefer to use the Antibody Diluent for membrane blocking and antibody incubation steps For blocking and antibody dilution for 50 membranes (1 L buffer 10 mL per membrane)