Learn More
Biotium Blocking Buffers Background Reducers Fluorescence Microscopy Western and Protein Analysis Chemical Reagents 1/EA
Catalog No. 5000501262
Encompass_Preferred
Bovine serum albumin (BSA) is commonly used as a stabilizing agent for proteins and enzymes including dilute solutions of antibody and use as a blocking agent to reduce non-specific binding It is also suitable for use in protease sensitive assays such as RIA and EIA as well as nucleic acid hybridization This BSA is IgG- and protease-free
Catalog No. 50-005-01262 Supplier Biotium Supplier No. 22013
Bovine serum albumin (BSA) is commonly used as a stabilizing agent for proteins and enzymes including dilute solutions of antibody and use as a blocking agent to reduce non-specific binding It is also suitable for use in protease sensitive assays such as RIA and EIA as well as nucleic acid hybridization This BSA is IgG- and protease-free