Learn More
Nacalai USA Inc BLOCKING ONE, 500ML
Catalog No. NC0589416
Encompass
Blocking One is superior to conventional blocking solutions. The preservative does not affect the enzyme activity of POD or ALP. Only simple refrigerator storage is necessary, even after opening the bottle.
Catalog No. NC0589416 Supplier Nacalai USA Inc Supplier No. 0395395
Blocking One is superior to conventional blocking solutions. The preservative does not affect the enzyme activity of POD or ALP. Only simple refrigerator storage is necessary, even after opening the bottle.