Learn More
Santa Cruz Biotechnology UltraCruz Blocking Reagent, 100 mL
Catalog No. NC1517984
Encompass
Plant protein derived universal antibody diluent and blocking reagent recommended for use in immunohistochemistry (IHC), immunofluorescence (IF), Western blotting (WB), and for other research applications. Animal-free alternative to sera, BSA or non-fat dry milk. Designed to stabilize diluted antibody solutions and minimize background due to non-specific binding in different antibody-based research applications. The UltraCruz Blocking Reagent is especially convenient in multiplex IHC (multiple antigen labeling) applications where antibodies from several different species are used.
Catalog No. NC1517984 Supplier Santa Cruz Biotechnology Supplier No. SC516214
Plant protein derived universal antibody diluent and blocking reagent recommended for use in immunohistochemistry (IHC), immunofluorescence (IF), Western blotting (WB), and for other research applications. Animal-free alternative to sera, BSA or non-fat dry milk. Designed to stabilize diluted antibody solutions and minimize background due to non-specific binding in different antibody-based research applications. The UltraCruz Blocking Reagent is especially convenient in multiplex IHC (multiple antigen labeling) applications where antibodies from several different species are used.