Learn More
Diagnostic Biosystems BLOCKING REAGENT Mouse on Mouse (MOM) Blocking Reagent
Catalog No. 502407390
Encompass_Preferred
Mouse on Mouse (MOM) Blocking Reagent is a unique serum-free blocking solution used to reduce nonspecific background staining from mouse antibodies. This blocking reagent has been shown to be more superior than serum-based blocking solutions.\n\nBlock nonspecific background staining observed during immunostaining using mouse antibodies on mouse tissue.
Catalog No. 50-240-7390 Supplier Diagnostic Biosystems Supplier No. K065
Mouse on Mouse (MOM) Blocking Reagent is a unique serum-free blocking solution used to reduce nonspecific background staining from mouse antibodies. This blocking reagent has been shown to be more superior than serum-based blocking solutions.\n\nBlock nonspecific background staining observed during immunostaining using mouse antibodies on mouse tissue.