Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Blocking Solution, 500 mL
SDP

Catalog No. 507517659
Encompass_Preferred

Blocking Solution minimizes non-specific binding, reduces background, and enhances assay sensitivity. Blocking is a very important step of Western blotting, as it prevents antibodies from binding to the membrane nonspecifically. Non-specific binding obscures the true signal, leading to inaccurate results.

Directions for use: Blocking Solution should be used undiluted. Use 0.1 ml of Blocking Solution per cm 2 of filter area. A detergent, e.g., Tween 20, may be added to further reduce nonspecific background.

Storage Conditions: -10 °C to - 20 °C
Shipping Condition: on dry ice
Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7659 Supplier Quality Biological Inc Supplier No. 351080101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blocking Solution minimizes non-specific binding, reduces background, and enhances assay sensitivity. Blocking is a very important step of Western blotting, as it prevents antibodies from binding to the membrane nonspecifically. Non-specific binding obscures the true signal, leading to inaccurate results.

Directions for use: Blocking Solution should be used undiluted. Use 0.1 ml of Blocking Solution per cm 2 of filter area. A detergent, e.g., Tween 20, may be added to further reduce nonspecific background.

Storage Conditions: -10 °C to - 20 °C
Shipping Condition: on dry ice
Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.