Learn More
Seracare Life Sciences Inc BLOCKING SOLUTION CONCENTRATE
Catalog No. 5067560
Encompass_Preferred
Blocking solution concentrate is used in immunohistochemistry applications where endogenous peroxidase activity causes non-specific signal and high background. Supplied as a 10X concentrate; working solution is prepared by diluting 1:10 in reagent quality water. Also available as part of HistoMark TrueBlue, HistoMark ORANGE and HistoMark BLACK substrate kits.
This item is not returnable. View return policy
Catalog No. 50-675-60 Supplier Seracare Life Sciences Inc Supplier No. 55600006
Blocking solution concentrate is used in immunohistochemistry applications where endogenous peroxidase activity causes non-specific signal and high background. Supplied as a 10X concentrate; working solution is prepared by diluting 1:10 in reagent quality water. Also available as part of HistoMark TrueBlue, HistoMark ORANGE and HistoMark BLACK substrate kits.