Learn More
Electron Microscopy Sciences Aurion Blocking Solution Donkey 30 ML
SDP
Catalog No. 5024769
Encompass_Preferred
Aurion has developed Blocking Solutions which effectively block ‘sticky' surfaces by multi-point hydrophobic and charge-based interactions. The Blocking Solutions are specifically designed to meet the characteristics of each type of secondary conjugate. With adequate blocking and incubation media, background reactions are minimized whereas specific reactions are not hampered.
Catalog No. 50-247-69 Supplier Electron Microscopy Sciences Supplier No. 25599
Aurion has developed Blocking Solutions which effectively block ‘sticky' surfaces by multi-point hydrophobic and charge-based interactions. The Blocking Solutions are specifically designed to meet the characteristics of each type of secondary conjugate. With adequate blocking and incubation media, background reactions are minimized whereas specific reactions are not hampered.