Learn More
Electron Microscopy Sciences Aurion Blocking Solution Rabbit 30 ML
SDP
Supplier:  Electron Microscopy Sciences 25597
Encompass_Preferred
Aurion has developed Blocking Solutions which effectively block ‘sticky' surfaces by multi-point hydrophobic and charge-based interactions. The Blocking Solutions are specifically designed to meet the characteristics of each type of secondary conjugate. With adequate blocking and incubation media, background reactions are minimized whereas specific reactions are not hampered.
Catalog No. 50-247-67