Learn More
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Catalog No. 88860120 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Places:
1
Catalog No. No. of Places
88-860-120 1
1 options
Catalog No. 88-860-120
Add to Cart
Add to Cart
Constructed of aluminum alloy
A variety of blocks for any application
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.2 mL
For Use With (Equipment) Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Material Aluminum Alloy
No. of Places 1
Type Block