Learn More
Selleck Chemical LLC Blonanserin S2112-20mg
SDP
Catalog No. 503108414
Encompass_Preferred
Blonanserin (AD 5423) is a relatively selective serotonin (5-HT)2A and dopamine D2 antagonist used the treatment of schizophrenia
Catalog No. 50-310-8414 Supplier Selleck Chemical LLC Supplier No. S211220MG
Blonanserin (AD 5423) is a relatively selective serotonin (5-HT)2A and dopamine D2 antagonist used the treatment of schizophrenia