Learn More
Cayman Chemical Blonanserin, Purity: ≥98%, CAS: 132810-10-7, 100 mg
SDP
Catalog No. 502969285
Encompass_Preferred
A mixed antagonist at dopamine D2, D3, and serotonin 5-HT2A receptors (Kis = 0.14, 0.49, and 0.81 nM, respectively) that also binds to 5-HT6 (Ki = 11.7 nM); developed as a novel antipsychotic that displays 20- and 94-fold higher affinity for D2 receptors compared to haloperidol and risperidone, respectively
Catalog No. 50-296-9285 Supplier Cayman Chemical Supplier No. 16480100 MG
A mixed antagonist at dopamine D2, D3, and serotonin 5-HT2A receptors (Kis = 0.14, 0.49, and 0.81 nM, respectively) that also binds to 5-HT6 (Ki = 11.7 nM); developed as a novel antipsychotic that displays 20- and 94-fold higher affinity for D2 receptors compared to haloperidol and risperidone, respectively