Learn More
Kol Biomedical Instruments Inc Blood Administration Set with 150 Micron Filter and 50 cc BD Syringe, 24/cs
SDP
Catalog No. NC1037824
Encompass
The Charter Medical Neonatal Syringe Set with 150-micron filter is designed to facilitate delivery of small aliquots of whole blood, cellular (red cells and platelets) components and fresh frozen plasma components for neonatal/pediatric transfusion.
Catalog No. NC1037824 Supplier Kol Biomedical Instruments Inc Supplier No. CM0396010F
The Charter Medical Neonatal Syringe Set with 150-micron filter is designed to facilitate delivery of small aliquots of whole blood, cellular (red cells and platelets) components and fresh frozen plasma components for neonatal/pediatric transfusion.