Learn More
NEOGEN Blood Agar Base, 10 kg
SDP
Catalog No. 502015374
Encompass_Preferred
Blood Agar Base is used with blood for the isolation and cultivation of a wide variety of fastidious microorganisms. Blood Agar Base is not intended for use in the diagnosis of disease or other conditions in humans. An inexpensive, general purpose agar base which, with the addition of 5-7% sterile blood, can be used to cultivate a wide range of microorganisms of indicative significance. Typical hemolysis patterns are obtained with this medium.  Formula:Beef Extract - 10.0 g/LBalanced Peptone - 10.0 g/LSodium Chloride - 5.0 g/LAgar - 12.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/blood-agar-base
Catalog No. 50-201-5374 Supplier NEOGEN Supplier No. 700004462
Blood Agar Base is used with blood for the isolation and cultivation of a wide variety of fastidious microorganisms. Blood Agar Base is not intended for use in the diagnosis of disease or other conditions in humans. An inexpensive, general purpose agar base which, with the addition of 5-7% sterile blood, can be used to cultivate a wide range of microorganisms of indicative significance. Typical hemolysis patterns are obtained with this medium.  Formula:Beef Extract - 10.0 g/LBalanced Peptone - 10.0 g/LSodium Chloride - 5.0 g/LAgar - 12.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/blood-agar-base