Learn More
Bioworld Blood Agar Base #2, 500 g
Catalog No. 50254277
Encompass_Preferred
Blood Agar Base #2Used for the isolation, cultivation, and detection of hemolytic activity of a variety of fastidious microorganisms Media is usually prepared with 5 to 10% bloodpH: 7.4Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: Plain – medium to dark amber agar With 5% sheep blood – cherry red agarSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Sodium Chloride Agar Liver Digest Yeast Extract
Catalog No. 50-254-277 Supplier Bioworld Supplier No. 306200101
Blood Agar Base #2Used for the isolation, cultivation, and detection of hemolytic activity of a variety of fastidious microorganisms Media is usually prepared with 5 to 10% bloodpH: 7.4Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: Plain – medium to dark amber agar With 5% sheep blood – cherry red agarSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Sodium Chloride Agar Liver Digest Yeast Extract