Learn More
MOLTOX Molecular Toxicology Inc Blood Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948805
Encompass_Preferred
Blood Agar is used to cultivate, isolate and detect hemolytic activity of streptococci and other fastidious microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-805 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S005
Blood Agar is used to cultivate, isolate and detect hemolytic activity of streptococci and other fastidious microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.