Description
Cultivate Haemophilus species with this ready prepared medium.
Cultivate Haemophilus species with Thermo Scientific™ Remel™ Blood Agar with 5% Rabbit Blood. Haemophilus spp. requires hemin (X factor) and NAD (V factor) for growth, both of which are supplied by Rabbit blood. Horse blood does not contain V factor -inactivating enzymes and supplies X and V factors, one or both of which are required for the growth of Haemophilus spp. Rabbit blood also allows for differentiation of hemolytic and nonhemolytic species of Haemophilus.
Members of the genus Haemophilus are fastidious Gram-negative bacilli that commonly cause infections of upper respiratory tract. Blood Agar with 5% Rabbit Blood can be used in qualitative procedures for the cultivation of Haemophilus species.
- Rabbit blood in the medium does not contain V factor -inactivating enzymes and supplies X and V factors, one or both of which are required for the growth of Haemophilus Rabbit blood also allows for differentiation of hemolytic and nonhemolytic species of Haemophilus.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Blood Agar (TSA w/5% Rabbit Blood) |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |