Learn More
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium
Catalog No. R01201 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
150/Pk.
20/Pk.
Format:
10mm x 100mm Spacesaver Plate
10mm x 85mm Spacesaver plate
12mm x 85mm Monoplate
15mm x 103mm Tube
85mm Monoplate
Tube
Includes:
TSA w/10% Sheep Blood
TSA w/5% Sheep Blood
Manufacturing Origin:
United States
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Includes Manufacturing Origin
R01201 15/Pk. 10mm x 85mm Spacesaver plate TSA w/5% Sheep Blood United States
R01200 10/Pk. 85mm Monoplate TSA w/5% Sheep Blood United States
R01202 100/Pk. 85mm Monoplate TSA w/5% Sheep Blood United States
R060312 20/Pk. Tube TSA w/5% Sheep Blood United States
R01204 10/Pk. 12mm x 85mm Monoplate TSA w/10% Sheep Blood United States
R01198 150/Pk. 10mm x 100mm Spacesaver Plate TSA w/5% Sheep Blood United States
R060310 100/Pk. 15mm x 103mm Tube TSA w/5% Sheep Blood United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. R01201 Supplier Thermo Scientific™ Supplier No. R01201
This item is not returnable. View return policy
Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote hemolysis.
Grow a wide variety of fastidious organisms on Thermo Scientific™ Blood Agar (TSA with Sheep Blood), a general purpose non-selective medium with the addition of sheep blood in the formulation to promote the presence and visualization of haemolytic reactions so it can be used as a diagnostic tool.
Tryptone Soya Agar (casein soya bean digest agar) with defibrinated sheep blood is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown either in deep cultures or by incubation under anaerobic conditions. The combination of casein digest and papaic digest of soybean meal leads to optimal growth due to synergy between the protein supply of casein and the carbohydrate supply of soybeans. Sterile defibrinated sheep blood used to enrich the medium produces the hemolysis characteristics of different bacteria, such as streptococci, listeria, hemolytic Staphylococcus, Escherichia coli and Pseudomonas.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM.
Format 10mm x 85mm Spacesaver plate
Manufacturing Origin United States
Media Type Blood Agar
Product Type Agar Plate
Includes TSA w/5% Sheep Blood
Quantity 15/Pk.